CAS RN 125-04-2
CAS RN 125-04-2
IUPAC Name:
sodium 4-{2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethoxy}-4-oxobutanoate
Synonyms:
sodium hydrocortisone 21-succinate
Molecular Weight (g/mol):
484.52
Molecular Formula:
C25H33NaO8
InChi Key:
HHZQLQREDATOBM-CODXZCKSSA-M
SMILES:
[Na+].C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@]2(O)C(=O)COC(=O)CCC([O-])=O
1
–
1
of
1
results
1
STEMCELL Technologies Hydrocortisone
Enhances proliferation and differentiation of endothelial, epithelial, mesenchymal, and oligodendrocyte cells