man

Test

CAS RN 1255517-77-1

CH3H3CNNHNNOCH3ONH2O

CAS RN 1255517-77-1

IUPAC Name: 2-cyclohexyl-6-methoxy-N-[1-(propan-2-yl)piperidin-4-yl]-7-[3-(pyrrolidin-1-yl)propoxy]quinazolin-4-amine hydrate
Synonyms: 2-cyclohexyl-N-(1-isopropylpiperidin-4-yl)-6-methoxy-7-[3-(pyrrolidin-1-yl)propoxy]quinazolin-4-amine hydrate
Molecular Weight (g/mol): 527.75
Molecular Formula: C30H49N5O3
InChi Key: LLJGACAJGYXBTL-UHFFFAOYSA-N
SMILES: O.COC1=CC2=C(NC3CCN(CC3)C(C)C)N=C(N=C2C=C1OCCCN1CCCC1)C1CCCCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results