man

Test

CAS RN 1255942-06-3

NOH2N

CAS RN 1255942-06-3

IUPAC Name: 3-amino-1-{2-azatricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl}propan-1-one
Synonyms: 3-amino-1-{2-azatricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl}propan-1-one
Molecular Weight (g/mol): 276.34
Molecular Formula: C18H16N2O
InChi Key: OCCYFTDHSHTFER-UHFFFAOYSA-N
SMILES: NCCC(=O)N1CC2=CC=CC=C2C#CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)