man

Test

CAS RN 1256360-47-0

BOOH3CCH3H3CH3CBrF

CAS RN 1256360-47-0

IUPAC Name: 2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[5-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 314.99
Molecular Formula: C13H17BBrFO2
InChi Key: UEBRMJPFPXLIMK-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=CC(CBr)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results