CAS RN 126-72-7
CAS RN 126-72-7
IUPAC Name:
tris(2,3-dibromopropyl) phosphate
Synonyms:
tris(2,3-dibromopropyl) phosphate
Molecular Weight (g/mol):
697.61
Molecular Formula:
C9H15Br6O4P
InChi Key:
PQYJRMFWJJONBO-UHFFFAOYNA-N
SMILES:
BrCC(Br)COP(=O)(OCC(Br)CBr)OCC(Br)CBr