accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1261571-92-9

H3COOClFFF

CAS RN 1261571-92-9

IUPAC Name: 3-methoxy-4-(trifluoromethyl)benzoyl chloride
Synonyms: 3-methoxy-4-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: RDTSCEWYAIWTQO-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(=C1)C(Cl)=O)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products