CAS RN 1261588-72-0
CAS RN 1261588-72-0
IUPAC Name:
ethyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Synonyms:
ethyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate
Molecular Weight (g/mol):
190.2
Molecular Formula:
C10H10N2O2
InChi Key:
CKCBQXQNAUHBDX-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C2C=CNC2=NC=C1
1
–
1
of
1
results
1
Ethyl 7-azaindole-4-carboxylate, 97%, Thermo Scientific Chemicals
CAS: 1261588-72-0 Molecular Formula: C10H10N2O2 Molecular Weight (g/mol): 190.202 MDL Number: MFCD15071133 InChI Key: CKCBQXQNAUHBDX-UHFFFAOYSA-N PubChem CID: 56773892 IUPAC Name: ethyl 1H-pyrrolo[2,3-b]pyridine-4-carboxylate SMILES: CCOC(=O)C1=C2C=CNC2=NC=C1