accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 12626-81-2

OOOOOTiZrPb

CAS RN 12626-81-2

IUPAC Name: titanium(4+) zirconium(4+) λ²-lead(2+) pentaoxidandiide
Synonyms: lead zirconate titanate
Molecular Weight (g/mol): 426.3
Molecular Formula: O5PbTiZr
InChi Key: HFGPZNIAWCZYJU-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[O--].[Ti+4].[Zr+4].[Pb++]
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products