CAS RN 1268524-71-5
CAS RN 1268524-71-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ (-)-JQ1

Inactive control for (+)-JQ1
2
Medchem Express (R)-(-)-JQ1 Enantiomer, MedChemExpress
MedChemExpress (R)-(-)-JQ1 Enantiomer is the stereoisomer of (+)-JQ1. (+)-JQ1 potently decreases expression of both BRD4 target genes, whereas (R)-(-)-JQ1 Enantiomer has no effect.

3