man

Test

CAS RN 127271-65-2

H3COFFFF

CAS RN 127271-65-2

IUPAC Name: 1-fluoro-4-methoxy-2-(trifluoromethyl)benzene
Synonyms: 1-fluoro-4-methoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: VUSCMOUYRPIURE-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results