accessibility menu, dialog, popup

UserName

CAS RN 127408-31-5

CH3ONOH(R)(S)HOH3CH3CON

CAS RN 127408-31-5

IUPAC Name: N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-yl]-N-hydroxyacetamide
Synonyms: N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl]-N-hydroxyacetamide
Molecular Weight (g/mol): 276.29
Molecular Formula: C14H16N2O4
InChi Key: IWTCFIIOUXJOOV-OLZOCXBDSA-N
SMILES: CC(=O)N(O)[C@H]1[C@H](O)C(C)(C)OC2=CC=C(C=C12)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results