accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 128562-25-4

OOOS

CAS RN 128562-25-4

IUPAC Name: 2-oxo-3-(thiophen-2-yl)propanoate
Synonyms: 2-oxo-3-(thiophen-2-yl)propanoate
Molecular Weight (g/mol): 169.17
Molecular Formula: C7H5O3S
InChi Key: DFEKJNVSDUKUIR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(=O)CC1=CC=CS1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products