man

Test

CAS RN 128639-02-1

H3COOClNNCH3NFFOFCl

CAS RN 128639-02-1

IUPAC Name: ethyl 2-chloro-3-{2-chloro-5-[4-(difluoromethyl)-3-methyl-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]-4-fluorophenyl}propanoate
Synonyms: carfentrazone-ethyl aurora 50WG aurora kuaimieling spotlight spotlight 24EC aurora (pesticide)
Molecular Weight (g/mol): 412.19
Molecular Formula: C15H14Cl2F3N3O3
InChi Key: MLKCGVHIFJBRCD-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Cl)CC1=CC(N2N=C(C)N(C(F)F)C2=O)=C(F)C=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results