man

Test

CAS RN 129825-11-2

H3COClFFF

CAS RN 129825-11-2

IUPAC Name: 1-[4-chloro-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-chloro-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 222.59
Molecular Formula: C9H6ClF3O
InChi Key: UYNMUXTXDHJBEN-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results