CAS RN 130152-35-1
CAS RN 130152-35-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ (+)-Igmesine hydrochloride
Selective sigma1 ligand; antidepressant