accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13037-60-0

BrNCS

CAS RN 13037-60-0

IUPAC Name: 1-bromo-2-isothiocyanatobenzene
Synonyms: 1-bromo-2-isothiocyanatobenzene
Molecular Weight (g/mol): 214.08
Molecular Formula: C7H4BrNS
InChi Key: PAFORXDSYWMYGP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products