accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13041-70-8

Br(E/Z)

CAS RN 13041-70-8

IUPAC Name: 1-bromo-4-(2-phenylethenyl)benzene
Synonyms: 1-bromo-4-(2-phenylethenyl)benzene
Molecular Weight (g/mol): 259.15
Molecular Formula: C14H11Br
InChi Key: ZZMMKLVIBZWGPK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=CC2=CC=CC=C2)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products