man

Test

CAS RN 1304778-15-1

CH3NNONNNO

CAS RN 1304778-15-1

IUPAC Name: 1-{1-[4-(1-benzofuran-2-yl)pyrimidin-2-yl]piperidin-3-yl}-3-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-{1-[4-(1-benzofuran-2-yl)pyrimidin-2-yl]piperidin-3-yl}-3-ethyl-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 439.52
Molecular Formula: C26H25N5O2
InChi Key: NXTBLPPTZRPJCA-UHFFFAOYNA-N
SMILES: CCN1C(=O)N(C2CCCN(C2)C2=NC=CC(=N2)C2=CC3=CC=CC=C3O2)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results