CAS RN 13100-46-4
CAS RN 13100-46-4
IUPAC Name:
4,5,6-tris(acetyloxy)-2-(hydroxymethyl)oxan-3-yl acetate
Synonyms:
4,5,6-tris(acetyloxy)-2-(hydroxymethyl)oxan-3-yl acetate
Molecular Weight (g/mol):
348.3
Molecular Formula:
C14H20O10
InChi Key:
FEQXFAYSNRWXDW-UHFFFAOYNA-N
SMILES:
CC(=O)OC1OC(CO)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals 1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose, 98%
CAS: 13100-46-4 Molecular Formula: C14H20O10 Molecular Weight (g/mol): 348.30 MDL Number: MFCD00063260 InChI Key: FEQXFAYSNRWXDW-UHFFFAOYNA-N Synonym: 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranose,beta-d-glucose 1,2,3,4-tetraacetate,2s,3r,4s,5r,6r-6-hydroxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate,2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-hydroxymethyl oxan-3-yl acetate,1,2,3,4-tetra-o-acetylglucopyranose,beta-d-glucopyranose 1,2,3,4-tetraacetate,1,2,3,4-tetra-o-acetyl-b-d-glucopyranose,beta-d-glucopyranose, 1,2,3,4-tetraacetate,b-d-glucopyranose,1,2,3,4-tetraacetate PubChem CID: 83136 IUPAC Name: [(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(hydroxymethyl)oxan-3-yl] acetate SMILES: CC(=O)OC1OC(CO)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O