CAS RN 13139-15-6
CAS RN 13139-15-6
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-4-methylpentanoic acid
Molecular Weight (g/mol):
231.29
Molecular Formula:
C11H21NO4
InChi Key:
MDXGYYOJGPFFJL-UHFFFAOYNA-N
SMILES:
CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
1
–
1
of
1
results
1
N-Boc-L-leucine hydrate, 99%, Thermo Scientific Chemicals
CAS: 13139-15-6 Molecular Formula: C11H21NO4 Molecular Weight (g/mol): 231.29 MDL Number: MFCD00066067 InChI Key: MDXGYYOJGPFFJL-SVGMAFHSNA-N Synonym: boc-leu-oh,boc-l-leucine,n-tert-butoxycarbonyl-l-leucine,n-boc-l-leucine,tert-butoxycarbonyl-l-leucine,tert-butoxycarbonylleucine,s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid,n-tert-butoxycarbonylleucine,boc-l-leu-oh,tert-butyloxycarbonyl-l-leucine PubChem CID: 83170 ChEBI: CHEBI:28702 IUPAC Name: (2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid SMILES: CC(C)C[C@H](NC(=O)OC(C)(C)C)C(O)=O