CAS RN 1315692-91-1
CAS RN 1315692-91-1
IUPAC Name:
2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms:
2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol):
255.07
Molecular Formula:
C12H16BF2NO2
InChi Key:
MQGLBRAIKSVSIT-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=C(F)C(N)=C(F)C=C1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 3-Amino-2,4-difluorobenzeneboronic acid pinacol ester, 96%
CAS: 1315692-91-1 Molecular Formula: C12H16BF2NO2 Molecular Weight (g/mol): 255.072 MDL Number: MFCD22419270 InChI Key: MQGLBRAIKSVSIT-UHFFFAOYSA-N Synonym: 3-amino-2,4-difluorobenzeneboronic acid pinacol ester,2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,3-amino-2,4-difluorobenzeneboronicacidpinacolester,2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline PubChem CID: 99738339 IUPAC Name: 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline SMILES: B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)N)F