man

Test

CAS RN 1317-34-6

OMnOMnO

CAS RN 1317-34-6

IUPAC Name: oxo[(oxomanganio)oxy]manganese
Synonyms: manganese(III) oxide
Molecular Weight (g/mol): 157.87
Molecular Formula: Mn2O3
InChi Key: GEYXPJBPASPPLI-UHFFFAOYSA-N
SMILES: O=[Mn]O[Mn]=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results