accessibility menu, dialog, popup

UserName

CAS RN 131747-41-6

NOHOFFF

CAS RN 131747-41-6

IUPAC Name: 2-(trifluoromethyl)pyridine-4-carboxylic acid
Synonyms: 2-(trifluoromethyl)pyridine-4-carboxylic acid
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: BZFGKBQHQJVAHS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results