man

Test

CAS RN 131747-54-1

NBrHO

CAS RN 131747-54-1

IUPAC Name: (2-bromopyridin-3-yl)methanol
Synonyms: (2-bromopyridin-3-yl)methanol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: QIQZUABMLGJKJP-UHFFFAOYSA-N
SMILES: OCC1=C(Br)N=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results