man

Test

CAS RN 131864-71-6

H3CClOH

CAS RN 131864-71-6

IUPAC Name: 1-(2-chlorophenyl)ethan-1-ol
Synonyms: 1-(2-chlorophenyl)ethanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: DDUBOVLGCYUYFX-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1Cl

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results