man

Test

CAS RN 13200-83-4

OHONO

CAS RN 13200-83-4

IUPAC Name: 1-formylpyrrolidine-2-carboxylic acid
Synonyms: 1-formylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 143.14
Molecular Formula: C6H9NO3
InChi Key: DHDRGOURKDLAOT-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1C=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results