man

Test

CAS RN 13222-85-0

CH3OOOH3C

CAS RN 13222-85-0

IUPAC Name: 4-methylbenzoyl 4-methylbenzoate
Synonyms: 4-methylbenzoyl 4-methylbenzoate
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: BJMLLSSSTGHJJE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results