man

Test

CAS RN 132606-40-7

CH3NClBr

CAS RN 132606-40-7

IUPAC Name: 3-bromo-6-chloro-2-methylpyridine
Synonyms: 3-bromo-6-chloro-2-methylpyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: DTFBHJWQTDQBEM-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(Cl)=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results