accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13327-27-0

CH3NNHO

CAS RN 13327-27-0

IUPAC Name: 6-methyl-2,3-dihydropyridazin-3-one
Synonyms: 6-methyl-2H-pyridazin-3-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: QZWIXLPWMGHDDD-UHFFFAOYSA-N
SMILES: CC1=NNC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products