CAS RN 133343-34-7
CAS RN 133343-34-7
IUPAC Name:
(2R)-2-acetamido-3-[(2R,3S,4R)-3-hydroxy-2-[(1S)-1-hydroxy-3-methylbutyl]-4-methyl-5-oxopyrrolidine-2-carbonylsulfanyl]propanoic acid
Synonyms:
lactacystin
Molecular Weight (g/mol):
390.45
Molecular Formula:
C16H26N2O7S
InChi Key:
KZZNIEYFCKAJPQ-VRKAVTRVSA-N
SMILES:
CC(C)C[C@H](O)[C@]1(NC(=O)[C@H](C)[C@@H]1O)C(=O)SC[C@H](NC(C)=O)C(O)=O