man

Test

CAS RN 1335243-55-4

H3COOCH3ICl

CAS RN 1335243-55-4

IUPAC Name: 2-chloro-3-iodo-1,4-dimethoxybenzene
Synonyms: 2-chloro-3-iodo-1,4-dimethoxybenzene
Molecular Weight (g/mol): 298.5
Molecular Formula: C8H8ClIO2
InChi Key: IMMMDYMSUZKGSS-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C(I)=C(OC)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results