CAS RN 133812-16-5
CAS RN 133812-16-5
IUPAC Name:
4-benzyl-3-[(2E)-but-2-enoyl]-1,3-oxazolidin-2-one
Synonyms:
4-benzyl-3-[(2E)-but-2-enoyl]-1,3-oxazolidin-2-one
Molecular Weight (g/mol):
245.28
Molecular Formula:
C14H15NO3
InChi Key:
UTZAFVPPWUIPBH-QHHAFSJGNA-N
SMILES:
C\C=C\C(=O)N1C(CC2=CC=CC=C2)COC1=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Locostatin
CAS: 133812-16-5 Molecular Formula: C14H15NO3 Molecular Weight (g/mol): 245.278 MDL Number: MFCD00278769 InChI Key: UTZAFVPPWUIPBH-QSLRECBCSA-N Synonym: locostatin,s-4-benzyl-3-but-2-enoyl oxazolidin-2-one,s,e-4-benzyl-3-but-2-enoyl oxazolidin-2-one,4s-n-crotonyl-4-benzyl-2-oxazolidinone,4s-3-e-but-2-enoyl-4-benzyl-2-oxazolidinone,4s-4-benzyl-3-e-but-2-enoyl-1,3-oxazolidin-2-one,cell sheet migration inhibitor, locostatin,4s-4-benzyl-3-2e-but-2-enoyl-1,3-oxazolidin-2-one,locostatin hplc,3-crotonyl-4beta-benzyloxazolidine-2-one PubChem CID: 5702600 IUPAC Name: (4S)-4-benzyl-3-[(E)-but-2-enoyl]-1,3-oxazolidin-2-one SMILES: CC=CC(=O)N1C(COC1=O)CC2=CC=CC=C2