CAS RN 133865-88-0
CAS RN 133865-88-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Ralfinamide, MedChemExpress
MedChemExpress Ralfinamide (FCE-26742A) is an orally available Na+ blocker derived from α-aminoamide, with function of suppressing pain.
