CAS RN 13389-42-9
CAS RN 13389-42-9
IUPAC Name:
(2E)-oct-2-ene
Synonyms:
2-octene (cis) 2-octene (trans)
Molecular Weight (g/mol):
112.22
Molecular Formula:
C8H16
InChi Key:
ILPBINAXDRFYPL-HWKANZROSA-N
SMILES:
CCCCC\C=C\C
1
–
2
of
2
results
1
trans-2-Octene, 97%, Thermo Scientific Chemicals
CAS: 13389-42-9 Molecular Formula: C8H16 Molecular Weight (g/mol): 112.22 MDL Number: MFCD00009532 InChI Key: ILPBINAXDRFYPL-HWKANZROSA-N Synonym: trans-2-octene,2-octene,e-2-octene,e-oct-2-ene,2-octene, e,2-octene, 2e,unii-g13g1yr8yw,oct-2-ene,g13g1yr8yw,octene-2 PubChem CID: 5364448 IUPAC Name: (E)-oct-2-ene SMILES: CCCCC\C=C\C
2
trans-2-Octene 98%, Thermo Scientific™
CAS: 13389-42-9 Molecular Formula: C8H16 Molecular Weight (g/mol): 112.22 MDL Number: MFCD00009532 InChI Key: ILPBINAXDRFYPL-HWKANZROSA-N Synonym: trans-2-octene,2-octene,e-2-octene,e-oct-2-ene,2-octene, e,2-octene, 2e,unii-g13g1yr8yw,oct-2-ene,g13g1yr8yw,octene-2 PubChem CID: 5364448 IUPAC Name: (2E)-oct-2-ene SMILES: CCCCC\C=C\C