man

Test

CAS RN 134031-24-6

NClOCl

CAS RN 134031-24-6

IUPAC Name: 2,4-dichloropyridine-3-carbaldehyde
Synonyms: 2,4-dichloropyridine-3-carbaldehyde
Molecular Weight (g/mol): 176
Molecular Formula: C6H3Cl2NO
InChi Key: GWBHJHIZHNTJLA-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C(Cl)=NC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results