man

Test

CAS RN 13446-30-5

H2OH2OH2OH2OH2OH2OMgOSSOO

CAS RN 13446-30-5

IUPAC Name: magnesium(2+) hexahydrate sulfanidesulfonate
Synonyms: magnesium(2+) hexahydrate thiosulphate
Molecular Weight (g/mol): 244.51
Molecular Formula: H12MgO9S2
InChi Key: CQDMJJVHDPDPHO-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Mg++].[O-]S([S-])(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results