man

Test

CAS RN 13453-38-8

H2OH2OH2OH2OH2OH2OTlONOOONOOONOO

CAS RN 13453-38-8

IUPAC Name: thallium(3+) hexahydrate trinitrate
Synonyms: thallium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 498.48
Molecular Formula: H12N3O15Tl
InChi Key: TWTZVACKNJEUQF-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Tl+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results