man

Test

CAS RN 13515-97-4

CH3OOH3CNH2HCl

CAS RN 13515-97-4

IUPAC Name: hydrogen methyl 2-aminopropanoate chloride
Synonyms: hydrogen methyl 2-aminopropanoate chloride
Molecular Weight (g/mol): 139.58
Molecular Formula: C4H10ClNO2
InChi Key: IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(C)N
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)