CAS RN 13551-73-0
CAS RN 13551-73-0
IUPAC Name:
5-chloro-1,3-dimethyl-4-nitro-1H-pyrazole
Synonyms:
5-chloro-1,3-dimethyl-4-nitropyrazole
Molecular Weight (g/mol):
175.57
Molecular Formula:
C5H6ClN3O2
InChi Key:
WBMBDRIOUHCVAS-UHFFFAOYSA-N
SMILES:
CN1N=C(C)C(=C1Cl)[N+]([O-])=O
1
–
1
of
1
results
1
5-Chloro-1,3-dimethyl-4-nitro-1H-pyrazole, 97%, Thermo Scientific™
CAS: 13551-73-0 Molecular Formula: C5H6ClN3O2 Molecular Weight (g/mol): 175.57 MDL Number: MFCD00052534 InChI Key: WBMBDRIOUHCVAS-UHFFFAOYSA-N Synonym: 5-chloro-1,3-dimethyl-4-nitro-1h-pyrazole,1,3-dimethyl-4-nitro-5-chloropyrazole,5-chloro-4-nitro-1,3-dimethylpyrazole,1,3-dimethyl-4-nitro-5-chloro-1h-pyrazole,1h-pyrazole,5-chloro-1,3-dimethyl-4-nitro,1h-pyrazole, 5-chloro-1,3-dimethyl-4-nitro,acmc-20adw4,buttpark 3711-59,5-chloro-1,3-dimethyl-4-nitro-1h-pyrazole #,5-chloro-4-nitro-1,3-dimethylpyrazole, PubChem CID: 279515 IUPAC Name: 5-chloro-1,3-dimethyl-4-nitropyrazole SMILES: CN1N=C(C)C(=C1Cl)[N+]([O-])=O