accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13566-03-5

PdOSOOO

CAS RN 13566-03-5

IUPAC Name: palladium(2+) sulfate
Synonyms: palladium(2+) sulfate
Molecular Weight (g/mol): 202.48
Molecular Formula: O4PdS
InChi Key: RFLFDJSIZCCYIP-UHFFFAOYSA-L
SMILES: [Pd++].[O-]S([O-])(=O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products