man

Test

CAS RN 13573-08-5

GeH4HIIH

CAS RN 13573-08-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 332.49
Molecular Formula: GeH6I2
InChi Key: PUZROLYCAWSHNL-UHFFFAOYSA-N
SMILES: [GeH4].I.I
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results