Target |
Histone Demethylase Inhibitors |
CAS |
1357302-64-7 |
Purity |
98% |
Chemical Name or Material |
4 '-((1 R,2 S)-2-aminocyclopropyl)biphenyl-3-ol |
Recommended Storage |
Store at -20°C |
Synonym |
4′-((1R,2S)-2-aminocyclopropyl)biphenyl-3-ol |
Molecular Formula |
C15H15NO |
Formula Weight |
225.29 |