CAS RN 1364917-14-5
CAS RN 1364917-14-5
IUPAC Name:
6-bromo-8-iodoimidazo[1,2-a]pyridine
Synonyms:
6-bromo-8-iodoimidazo[1,2-a]pyridine
Molecular Weight (g/mol):
322.93
Molecular Formula:
C7H4BrIN2
InChi Key:
OKEMQQRZPHOOGS-UHFFFAOYSA-N
SMILES:
BrC1=CN2C=CN=C2C(I)=C1
1
–
1
of
1
results
1
6-Bromo-8-iodoimidazo[1,2-a]pyridine, 95%, Thermo Scientific™
CAS: 1364917-14-5 Molecular Formula: C7H4BrIN2 Molecular Weight (g/mol): 322.931 MDL Number: MFCD21609484 InChI Key: OKEMQQRZPHOOGS-UHFFFAOYSA-N PubChem CID: 71299170 IUPAC Name: 6-bromo-8-iodoimidazo[1,2-a]pyridine SMILES: C1=CN2C=C(C=C(C2=N1)I)Br