accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1365531-82-3

CH3PH3CPCH3CH3OH3COH3COCH3OCH3(?)

CAS RN 1365531-82-3

IUPAC Name: {2'-[bis(4-methylphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(4-methylphenyl)phosphane
Synonyms: {2'-[bis(4-methylphenyl)phosphanyl]-4,4',6,6'-tetramethoxy-[1,1'-biphenyl]-2-yl}bis(4-methylphenyl)phosphane
Molecular Weight (g/mol): 698.78
Molecular Formula: C44H44O4P2
InChi Key: JXFDRZQHVPTIMH-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C(=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1)C1=C(OC)C=C(OC)C=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products