accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 136834-22-5

N(R)NONH(R)N(R)NO(R)OOOCH3OH3CPONCH3CH3H3CCH3NF

CAS RN 136834-22-5

IUPAC Name: N-[9-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-({[bis(propan-2-yl)amino](2-cyanoethoxy)phosphanyl}oxy)-3-fluorooxolan-2-yl)-9H-purin-6-yl]benzamide
Synonyms: N-[9-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-{[(2-cyanoethoxy)(diisopropylamino)phosphanyl]oxy}-3-fluorooxolan-2-yl)purin-6-yl]benzamide
Molecular Weight (g/mol): 875.94
Molecular Formula: C47H51FN7O7P
InChi Key: VCCMVPDSLHFCBB-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(OCC1OC(C(F)C1OP(OCCC#N)N(C(C)C)C(C)C)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products