accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13689-20-8

OP

CAS RN 13689-20-8

IUPAC Name: [cyclohexyl(phenyl)phosphoroso]benzene
Synonyms: [cyclohexyl(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 284.34
Molecular Formula: C18H21OP
InChi Key: ICVUZKQDJNUMKC-UHFFFAOYSA-N
SMILES: O=P(C1CCCCC1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products