CAS RN 137-17-7
CAS RN 137-17-7
IUPAC Namn:
2,4,5-trimethylaniline
Synonymer:
2,4,5-trimethylaniline
Molekylvikt (g/mol):
135.21
Molekylär sammansättning:
C9H13N
InChi Key:
BMIPMKQAAJKBKP-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(N)C=C1C
1
–
2
av
2
Resultat
1
SN/T 3045-2011 Azo Dyes Mixture 120 300 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality