accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 13701-64-9

CaCaCaOBOOOBOO

CAS RN 13701-64-9

IUPAC Name: tricalcium diborate
Synonyms: calcium borate
Molecular Weight (g/mol): 237.85
Molecular Formula: B2Ca3O6
InChi Key: VLCLHFYFMCKBRP-UHFFFAOYSA-N
SMILES: [Ca++].[Ca++].[Ca++].[O-]B([O-])[O-].[O-]B([O-])[O-]
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products