man

Test

CAS RN 137049-00-4

CH3NNSOOCl

CAS RN 137049-00-4

IUPAC Name: 1-methyl-1H-imidazole-4-sulfonyl chloride
Synonyms: 1-methylimidazole-4-sulfonyl chloride
Molecular Weight (g/mol): 180.61
Molecular Formula: C4H5ClN2O2S
InChi Key: KXUGUWTUFUWYRS-UHFFFAOYSA-N
SMILES: CN1C=NC(=C1)S(Cl)(=O)=O
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)