CAS RN 1373268-67-7
CAS RN 1373268-67-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express USL311, MedChemExpress
MedChemExpress USL311 is a selective CXCR4 antagonist, with anti-tumor activity. USL311 prevents the binding of stromal-cell derived factor-1 (SDF-1 or CXCL12) to CXCR4.
